Assay Details
| Gene Symbol: Dpy19l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166207.1 | NP_001159679.1 | 1 - 2 | 511 | 133 | |
| GenBank mRNA | AK030128.1 | - | 1 - 2 | 501 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpy19l2
Genomic Map