Assay Details
| Gene Symbol: Cib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019686.5 | NP_062660.1 | 2 - 3 | 140 | 68 | |
| GenBank mRNA | AB016080.1 | - | 2 - 3 | 140 | 68 | |
| AK005347.1 | - | 2 - 3 | 244 | 68 | ||
| AK013240.1 | - | 249 | 68 | |||
| BC005739.1 | - | 2 - 3 | 182 | 3599342 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cib2
Genomic Map