Assay Details
| Gene Symbol: Ddx25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013932.4 | NP_038960.2 | 10 - 11 | 1272 | 78 | |
| GenBank mRNA | AF142630.3 | - | 10 - 11 | 1203 | 78 | |
| AK050693.1 | - | 10 - 11 | 1267 | 78 | ||
| AK078340.1 | - | 10 - 11 | 1231 | 78 | ||
| BC024852.1 | - | 10 - 11 | 1216 | 5358404 | 78 | |
| BC061130.1 | - | 10 - 11 | 1227 | 6774579 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx25
Genomic Map