Assay Details
Gene Symbol: Ddx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013932.4 | NP_038960.2 | 6 - 7 | 586 | 66 | |
GenBank mRNA | AF142630.3 | - | 6 - 7 | 517 | 66 | |
AK050693.1 | - | 6 - 7 | 581 | 66 | ||
AK078340.1 | - | 6 - 7 | 545 | 66 | ||
BC024852.1 | - | 6 - 7 | 530 | 5358404 | 66 | |
BC061130.1 | - | 6 - 7 | 541 | 6774579 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
