Assay Details
| Gene Symbol: Ddx25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013932.4 | NP_038960.2 | 9 - 10 | 1111 | 79 | |
| GenBank mRNA | AF142630.3 | - | 9 - 10 | 1042 | 79 | |
| AK050693.1 | - | 9 - 10 | 1106 | 79 | ||
| AK078340.1 | - | 9 - 10 | 1070 | 79 | ||
| BC024852.1 | - | 9 - 10 | 1055 | 5358404 | 79 | |
| BC061130.1 | - | 9 - 10 | 1066 | 6774579 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx25
Genomic Map