Assay Details
Gene Symbol: Fam96a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026635.3 | NP_080911.1 | 4 - 5 | 625 | 97 | |
GenBank mRNA | AK002690.1 | - | 4 - 5 | 567 | 97 | |
AK003870.1 | - | 4 - 5 | 521 | 97 | ||
AK077700.1 | - | 4 - 5 | 626 | 97 | ||
AK170445.1 | - | 4 - 5 | 419 | 97 | ||
AK184756.1 | - | 2 - 3 | 74 | 97 | ||
BC005745.1 | - | 4 - 5 | 542 | 3600028 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327530 | BC005745.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327530 | AK002690.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327530 | AK003870.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327530 | AK077700.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327530 | AK170445.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327530 | BC005745.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327530 | AK002690.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327530 | AK003870.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327530 | AK077700.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P327530 | AK170445.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044297 | BC005745.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044297 | BE655498.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044297 | AK002690.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044297 | AK003870.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044297 | AK077700.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491860 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393467 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782996 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317652 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531262 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259850 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22141 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175182 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111858 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327530 | BC005745.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327530 | AK002690.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327530 | AK003870.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327530 | AK077700.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327530 | AK170445.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327530 | BC005745.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327530 | AK002690.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327530 | AK003870.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327530 | AK077700.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P327530 | AK170445.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044297 | BC005745.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044297 | BE655498.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044297 | AK002690.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044297 | AK003870.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044297 | AK077700.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591360 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591362 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591349 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591344 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591358 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591347 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591345 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591363 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591337 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591351 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591352 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591364 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591341 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591346 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591355 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26591354 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06823691 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111858 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175182 | - | - |
Whole Mouse Genome G4122A | A_51_P327530 | BC005745.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327530 | AK002690.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327530 | AK003870.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327530 | AK077700.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327530 | AK170445.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327530 | BC005745.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327530 | AK002690.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327530 | AK003870.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327530 | AK077700.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P327530 | AK170445.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111858 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175182 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08144393 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08144399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26591308 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07895581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07585981 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591344 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591367 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591358 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591364 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07673919 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07452422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591365 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07542248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591354 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591341 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08144393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591353 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591333 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591335 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591352 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591347 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591380 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07452427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26591351 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10586486 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10586488 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10586490 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10586485 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10586489 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10586487 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416634_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416633_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10586484 | - | - |
Mouse Genome 430 2.0 Array | 1416634_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1416634_at | - | - |
Mouse Genome 430 2.0 Array | 1416633_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1416633_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68250_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68250_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900006624.mm.1 | NM_026635.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900006624.mm.1 | NM_026635.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900006624.mm.1 | TC0900000863.mm.1 | MTA TC |
Mouse Expression Set 430 | 1416634_PM_at | - | - |
Mouse Expression Set 430 | 1416633_PM_a_at | - | - |
Mouse Expression Set 430 | 1416633_a_at | - | - |
Mouse Expression Set 430 | 1416634_at | - | - |
Murine Genome U74v2 Set | 96613_at | - | - |
Murine Genome U74v2 Set | 96613_PM_at | - | - |
Murine Genome U74v2 Set | 168871_PM_i_at | - | - |
Murine Genome U74v2 Set | 168871_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1416634_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416634_at | - | - |
Mouse Genome 430A 2.0 Array | 1416633_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1416633_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416634_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416633_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416633_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416634_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 917833 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3440082 | - | - |
MouseWG-6 v2 Expression BeadChip | 2510035 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2510035 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3440082 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002510035 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003440082 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600218 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2695450 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2695450 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2600218 | - | - |
mouse-6 v1.1 (Illumina) | scl0068250.1_125-S | - | - |
mouse-6 v1.1 (Illumina) | scl068250.6_215-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2510035 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3440082 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0068250.1_125-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl068250.6_215-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2510035 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440082 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2695450 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2600218 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600218 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2695450 | - | - |
