Assay Details
Gene Symbol: Ccdc97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028771.2 | NP_083047.1 | 2 - 3 | 595 | 53 | |
GenBank mRNA | AK004762.1 | - | 2 - 3 | 596 | 53 | |
AK013304.1 | - | 2 - 3 | 546 | 53 | ||
AK045509.1 | - | 2 - 3 | 587 | 53 | ||
AK088221.1 | - | 2 - 3 | 586 | 53 | ||
AK155466.1 | - | 2 - 3 | 578 | 53 | ||
BC006752.1 | - | 1 - 2 | 100 | 53 | ||
BC078634.1 | - | 2 - 3 | 562 | 5388281 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
