Assay Details
| Gene Symbol: Cyp2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028775.3 | NP_083051.1 | 7 - 8 | 1194 | 59 | |
| GenBank mRNA | AK004699.2 | - | 7 - 8 | 1194 | 59 | |
| AK087069.1 | - | 7 - 8 | 1182 | 59 | ||
| AK162947.1 | - | 7 - 8 | 1578 | 59 | ||
| BC034202.1 | - | 4 - 5 | 634 | 59 | ||
| BC064733.1 | - | 7 - 8 | 1211 | 30459121 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2s1
Genomic Map