Assay Details
| Gene Symbol: Bckdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007533.5 | NP_031559.3 | 3 - 4 | 401 | 67 | |
| GenBank mRNA | AK002435.1 | - | 3 - 4 | 389 | 67 | |
| AK154734.1 | - | 3 - 4 | 391 | 67 | ||
| BC003787.1 | - | 3 - 4 | 413 | 3600452 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bckdha
Genomic Map