Assay Details
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016876.3 | NP_058572.2 | 1 - 2 | 51 | 109 | |
GenBank mRNA | AF108214.1 | - | 1 - 2 | 65 | 109 | |
AK038249.1 | - | 1 - 2 | 61 | 109 | ||
AK168362.1 | - | 1 - 2 | 54 | 109 | ||
BC008511.1 | - | 1 - 2 | 51 | 3592537 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF108214.1, AK038249.1, AK168362.1, BC008511.1, NM_016876.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
