Assay Details
Gene Symbol: Dyx1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026314.3 | NP_080590.3 | 2 - 3 | 357 | 94 | |
GenBank mRNA | AK076862.1 | - | 2 - 3 | 322 | 94 | |
AK161368.1 | - | 2 - 3 | 322 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
