Assay Details
Gene Symbol: Dyx1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026314.3 | NP_080590.3 | 3 - 4 | 489 | 116 | |
GenBank mRNA | AY428507.1 | - | 3 - 4 | 489 | 116 | |
BC026462.1 | - | 3 - 4 | 446 | 4483015 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
