Assay Details
| Gene Symbol: Rinl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177158.5 | NP_796132.2 | 10 - 11 | 1550 | 76 | |
| GenBank mRNA | AK030993.1 | - | 10 - 11 | 1543 | 76 | |
| AK082828.1 | - | 3 - 4 | 786 | 76 | ||
| AK162469.1 | - | 3 - 4 | 455 | 76 | ||
| AK168472.1 | - | 3 - 4 | 713 | 76 | ||
| BC044761.1 | - | 10 - 11 | 1586 | 76 | ||
| BC044878.1 | - | 1 - 2 | 267 | 76 | ||
| BC046530.1 | - | 10 - 11 | 1586 | 5329601 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rinl
Genomic Map