Assay Details
Gene Symbol: Lgals4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010706.2 | NP_034836.1 | 5 - 6 | 899 | 64 | |
GenBank mRNA | AF510729.1 | - | 5 - 6 | 548 | 64 | |
AK145734.1 | - | 5 - 6 | 719 | 64 | ||
AY044870.1 | - | 5 - 6 | 571 | 64 | ||
BC011236.1 | - | 5 - 6 | 537 | 4205669 | 64 | |
BC021632.1 | - | 5 - 6 | 555 | 4218531 | 64 | |
BC030297.1 | - | 5 - 6 | 712 | 4986324 | 64 | |
BC094008.1 | - | 5 - 6 | 550 | 4986911 | 64 | |
BC141106.1 | - | 5 - 6 | 621 | 9055718 | 64 | |
BC145387.1 | - | 5 - 6 | 621 | 9053945 | 64 | |
EF017938.1 | - | 5 - 6 | 899 | 64 | ||
EF017939.1 | - | 5 - 6 | 899 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
