Assay Details
| Gene Symbol: Papl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175319.4 | NP_780528.1 | 12 - 13 | 1555 | 66 | |
| GenBank mRNA | AK049131.1 | - | 12 - 13 | 1556 | 66 | |
| BC094908.1 | - | 12 - 13 | 1451 | 30021730 | 66 | |
| BC132375.1 | - | 12 - 13 | 1542 | 40130652 | 66 | |
| BC138307.1 | - | 12 - 13 | 1541 | 8861327 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25022914 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25022916 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25022915 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25022917 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25022913 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137800 | BC094908.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137800 | BC132375.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137800 | BC138307.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137800 | NM_175319.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137800 | AK049131.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137800 | BC094908.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137800 | BC132375.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137800 | BC138307.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137800 | NM_175319.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137800 | AK049131.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651529 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595550 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346401 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28903 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770592 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150275 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459071 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94537 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122660 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116906 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165654 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137800 | BC094908.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137800 | BC132375.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137800 | BC138307.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137800 | NM_175319.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137800 | AK049131.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137800 | BC094908.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137800 | BC132375.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137800 | BC138307.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137800 | NM_175319.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137800 | AK049131.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022914 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122660 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165654 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116906 | - | - |
| Whole Mouse Genome G4122A | A_51_P137800 | BC094908.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137800 | BC132375.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137800 | BC138307.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137800 | NM_175319.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137800 | AK049131.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137800 | BC094908.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137800 | BC132375.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137800 | BC138307.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137800 | NM_175319.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137800 | AK049131.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113166 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100236 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165654 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122660 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116906 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05976679 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05904984 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05976681 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06289480 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05904988 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25022914 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25022916 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25022915 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25022917 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25022913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06335522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06448705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06277806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06658654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05988292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05904988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06448703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06650604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06335521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05976681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06142384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06404197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06319319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05904982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06448706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06448700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06289480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25022916 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561500 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561511 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561502 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561501 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561504 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561505 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440887_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561498 | - | - |
| Mouse Genome 430 2.0 Array | 1440887_at | - | - |
| Mouse Genome 430 2.0 Array | 1440887_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101744_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101744_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700023678.mm.1 | TC0700002703.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700023678.mm.1 | NM_175319.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023678.mm.1 | NM_175319.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023677.mm.1 | TC0700002703.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700023677.mm.1 | NM_175319.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700023677.mm.1 | NM_175319.4 | Refseq |
| Mouse Expression Set 430 | 1440887_at | - | - |
| Mouse Expression Set 430 | 1440887_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440887_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440887_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 802529 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2627488 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2627488 | - | - |
| mouse-6 v1.1 (Illumina) | scl31574.14.1_31-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6180133 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31574.14.1_31-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6180133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2938973 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2938973 | - | - |

Papl
Genomic Map