Assay Details
| Gene Symbol: Tmem25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027865.2 | NP_082141.1 | 6 - 7 | 899 | 65 | |
| GenBank mRNA | AK002841.1 | - | 6 - 7 | 900 | 65 | |
| AK090375.1 | - | 4 - 5 | 1173 | 65 | ||
| AK154206.1 | - | 5 - 6 | 1385 | 65 | ||
| AK165199.1 | - | 6 - 7 | 931 | 65 | ||
| AK165827.1 | - | 6 - 7 | 884 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem25
Genomic Map