Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900003719.mm.1 |
NM_001013373.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003719.mm.1 |
NM_001013373.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003719.mm.1 |
TC0900000539.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003718.mm.1 |
NM_001013373.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003718.mm.1 |
NM_001013373.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003718.mm.1 |
TC0900000539.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003720.mm.1 |
NM_001013373.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003720.mm.1 |
NM_001013373.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003720.mm.1 |
TC0900000539.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003722.mm.1 |
NM_001013373.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003722.mm.1 |
NM_001013373.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003722.mm.1 |
TC0900000539.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003721.mm.1 |
NM_001013373.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003721.mm.1 |
NM_001013373.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003721.mm.1 |
TC0900000539.mm.1 |
MTA TC |