Assay Details
| Gene Symbol: C2cd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081314.2 | NP_001074783.1 | 1 - 2 | 84 | 52 | |
| GenBank mRNA | BC039571.1 | - | 1 - 2 | 83 | 1515262 | 52 |
| BP769160.1 | - | 1 - 2 | 109 | 52 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC039571.1, NM_001081314.2 transcripts.
Set Membership: |
|

C2cd4b
Genomic Map