Assay Details
Gene Symbol: 4930533K18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015956.1 | - | 789 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2230823 | AK017042.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P149267 | AK015956.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P149267 | AK015956.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175281 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140213 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140213 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07100332 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07100240 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02154020 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07100267 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175281 | - | - |
Whole Mouse Genome G4122A | A_52_P305841 | AK017042.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112439 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175281 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08759548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08688528 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08958229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232301 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08770559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232320 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232137 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232208 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232236 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232154 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08866400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232305 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08866392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08317707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08317715 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232284 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232327 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232307 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232309 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08958220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08366924 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08571724 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08958222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232136 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08394122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08889108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232318 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232242 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08752569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232173 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232322 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232142 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08834009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08419907 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08925116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232279 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08394129 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08571717 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08219575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232325 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08413991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232308 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232330 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232192 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08419903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232277 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27232141 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1432172_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432148_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432148_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1436029_at | - | - |
Mouse Genome 430 2.0 Array | 1432148_at | - | - |
Mouse Genome 430 2.0 Array | 1432172_at | - | - |
Mouse Genome 430 2.0 Array | 1432172_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1436029_PM_at | - | - |
Mouse Expression Set 430 | 1423484_PM_at | - | - |
Mouse Expression Set 430 | 1423484_at | - | - |
Mouse Expression Set 430 | 1432148_at | - | - |
Mouse Expression Set 430 | 1432148_PM_at | - | - |
Mouse Expression Set 430 | 1436029_PM_at | - | - |
Mouse Expression Set 430 | 1432172_PM_at | - | - |
Mouse Expression Set 430 | 1436029_at | - | - |
Mouse Expression Set 430 | 1432172_at | - | - |
Mouse Genome 430A 2.0 Array | 1423484_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1423484_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432172_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432172_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432148_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432148_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 102120537 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102120537 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102120537 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233101 | - | - |
