Assay Details
| Gene Symbol: Bmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007555.3 | NP_031581.2 | 3 - 4 | 1565 | 75 | |
| GenBank mRNA | AK033362.1 | - | 3 - 4 | 1568 | 75 | |
| AK160412.1 | - | 673 | 75 | |||
| BC100751.1 | - | 3 - 4 | 830 | 75 | ||
| BC100752.1 | - | 3 - 4 | 830 | 75 | ||
| BC100753.1 | - | 3 - 4 | 836 | 75 | ||
| BC100754.1 | - | 3 - 4 | 830 | 75 | ||
| BC141283.1 | - | 3 - 4 | 1362 | 9055895 | 75 | |
| L41145.1 | - | 3 - 4 | 831 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmp5
Genomic Map