Assay Details
| Gene Symbol: Ccdc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166282.1 | NP_001159754.1 | 4 - 5 | 1001 | 70 | |
| NM_029212.3 | NP_083488.1 | 2 - 3 | 470 | 70 | ||
| GenBank mRNA | AB048860.1 | - | 2 - 3 | 544 | 70 | |
| AK015980.1 | - | 2 - 3 | 544 | 70 | ||
| BC116291.1 | - | 2 - 3 | 470 | 40094912 | 70 | |
| BC116292.1 | - | 2 - 3 | 470 | 40094918 | 70 | |
More Information
Set Membership: |
|

Ccdc33
Genomic Map