Assay Details
| Gene Symbol: Stra6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162475.1 | NP_001155947.1 | 5 - 6 | 485 | 65 | |
| NM_001162476.1 | NP_001155948.1 | 5 - 6 | 796 | 65 | ||
| NM_001162479.1 | NP_001155951.1 | 5 - 6 | 590 | 65 | ||
| NM_009291.2 | NP_033317.2 | 5 - 6 | 755 | 65 | ||
| GenBank mRNA | AF062476.1 | - | 5 - 6 | 755 | 65 | |
| BC075657.1 | - | 5 - 6 | 485 | 30605532 | 65 | |
| CF536552.1 | - | 5 - 6 | 574 | 65 | ||
| CF742607.1 | - | 5 - 6 | 589 | 65 | ||
| CX237908.1 | - | 3 - 4 | 282 | 65 | ||
More Information
Set Membership: |
|

Stra6
Genomic Map