Assay Details
| Gene Symbol: 6030419C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176921.3 | NP_795895.1 | 1 - 2 | 639 | 74 | |
| GenBank mRNA | AK031382.1 | - | 1 - 2 | 641 | 74 | |
| AK047220.1 | - | 1 - 2 | 280 | 74 | ||
More Information
Additional Information:
				    	 
					    	
					        	
					            	This assay is in the 5'UTR region of the AK031382.1, NM_176921.3 transcripts.
					        	
					    	            
				    	
			    	
			    
			Set Membership: | 
		   
					
						
							 | 
				

6030419C18Rik
Genomic Map