Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900004088.mm.1 |
NM_172921.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004088.mm.1 |
NM_172921.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004088.mm.1 |
TC0900000581.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004085.mm.1 |
TC0900000581.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004089.mm.1 |
NM_172921.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004089.mm.1 |
NM_172921.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004089.mm.1 |
TC0900000581.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004090.mm.1 |
NM_172921.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004090.mm.1 |
NM_172921.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004090.mm.1 |
TC0900000581.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004092.mm.1 |
NM_172921.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004092.mm.1 |
NM_172921.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004092.mm.1 |
TC0900000581.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004091.mm.1 |
NM_172921.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004091.mm.1 |
NM_172921.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004091.mm.1 |
TC0900000581.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004087.mm.1 |
TC0900000581.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004094.mm.1 |
NM_172921.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004094.mm.1 |
NM_172921.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004094.mm.1 |
TC0900000581.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004080.mm.1 |
TC0900000581.mm.1 |
MTA TC |