Assay Details
| Gene Symbol: Opcml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177906.4 | NP_808574.2 | 5 - 6 | 760 | 93 | |
| GenBank mRNA | AK158538.1 | - | 5 - 6 | 1082 | 93 | |
| BC076581.1 | - | 5 - 6 | 760 | 30542711 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Opcml
Genomic Map