Assay Details
| Gene Symbol: Glb1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153803.1 | NP_722498.1 | 3 - 4 | 372 | 80 | |
| GenBank mRNA | AK136711.1 | - | 3 - 4 | 406 | 80 | |
| AK143057.1 | - | 2 - 3 | 303 | 80 | ||
| BC038479.1 | - | 3 - 4 | 372 | 4503030 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glb1l2
Genomic Map