Assay Details
| Gene Symbol: Glb1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153803.1 | NP_722498.1 | 8 - 9 | 821 | 69 | |
| GenBank mRNA | AK033400.1 | - | 4 - 5 | 404 | 69 | |
| AK136711.1 | - | 8 - 9 | 855 | 69 | ||
| AK143057.1 | - | 7 - 8 | 752 | 69 | ||
| BC038479.1 | - | 8 - 9 | 821 | 4503030 | 69 | |
More Information
Set Membership: |
|

Glb1l2
Genomic Map