Assay Details
| Gene Symbol: Serinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019760.3 | NP_062734.1 | 4 - 5 | 591 | 131 | |
| GenBank mRNA | AB029500.1 | - | 4 - 5 | 535 | 131 | |
| AF181685.1 | - | 4 - 5 | 511 | 131 | ||
| AK002847.1 | - | 4 - 5 | 540 | 131 | ||
| AK005203.1 | - | 4 - 5 | 535 | 131 | ||
| AK088340.1 | - | 4 - 5 | 533 | 131 | ||
| AK133668.1 | - | 4 - 5 | 532 | 131 | ||
| AK133975.1 | - | 4 - 5 | 532 | 131 | ||
| AK135359.1 | - | 4 - 5 | 509 | 131 | ||
| AK141574.1 | - | 4 - 5 | 540 | 131 | ||
| AK148745.1 | - | 4 - 5 | 512 | 131 | ||
| AK159280.1 | - | 4 - 5 | 506 | 131 | ||
| AK159436.1 | - | 4 - 5 | 558 | 131 | ||
| AK167817.1 | - | 4 - 5 | 539 | 131 | ||
| BC006940.1 | - | 4 - 5 | 564 | 131 | ||
| BC017148.1 | - | 4 - 5 | 481 | 4506673 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serinc1
Genomic Map