Assay Details
| Gene Symbol: Smpdl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020561.2 | NP_065586.3 | 5 - 6 | 868 | 111 | |
| GenBank mRNA | AK151683.1 | - | 5 - 6 | 754 | 111 | |
| AK152282.1 | - | 5 - 6 | 769 | 111 | ||
| AK196251.1 | - | 3 - 4 | 340 | 111 | ||
| BC139288.1 | - | 5 - 6 | 760 | 8862310 | 111 | |
| BC139289.1 | - | 5 - 6 | 760 | 8862311 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smpdl3a
Genomic Map