Assay Details
| Gene Symbol: Csf3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007782.3 | NP_031808.2 | 8 - 9 | 1260 | 117 | |
| NR_045561.1 | 7 - 8 | 1161 | 117 | |||
| GenBank mRNA | AK089305.1 | - | 8 - 9 | 1205 | 117 | |
| AK144318.1 | - | 7 - 8 | 1161 | 117 | ||
| BC116635.1 | - | 7 - 8 | 1137 | 40081333 | 117 | |
| M58288.1 | - | 8 - 9 | 1183 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csf3r
Genomic Map