Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400029007.mm.1 |
TC0400003559.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029007.mm.1 |
NM_026670.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029007.mm.1 |
NM_026670.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029008.mm.1 |
TC0400003559.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029008.mm.1 |
NM_026670.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029008.mm.1 |
NM_026670.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029010.mm.1 |
TC0400003559.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400029010.mm.1 |
NM_026670.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029010.mm.1 |
NM_026670.4 |
Refseq |