Assay Details
| Gene Symbol: Zmym1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026670.4 | NP_080946.3 | 6 - 7 | 981 | 67 | |
| GenBank mRNA | AK016783.1 | - | 6 - 7 | 977 | 67 | |
| AK035618.1 | - | 6 - 7 | 981 | 67 | ||
| AK049568.1 | - | 6 - 7 | 971 | 67 | ||
| AK132399.1 | - | 2 - 3 | 1943 | 67 | ||
| AK166769.1 | - | 6 - 7 | 964 | 67 | ||
| AK167508.1 | - | 6 - 7 | 987 | 67 | ||
| BC027750.1 | - | 6 - 7 | 1008 | 67 | ||
| BC043454.1 | - | 5 - 6 | 598 | 67 | ||
| BC064774.1 | - | 3 - 4 | 345 | 30461626 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC064774.1 transcripts.
Set Membership: |
|

Zmym1
Genomic Map