Assay Details
| Gene Symbol: Mcm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027830.2 | NP_082106.2 | 9 - 10 | 1795 | 72 | |
| GenBank mRNA | AK133212.1 | - | 2 - 3 | 208 | 72 | |
| AK133345.1 | - | 2 - 3 | 1022 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133212.1 transcripts.
Set Membership: |
|

Mcm9
Genomic Map