Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000015182.mm.1 |
NM_001204983.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015182.mm.1 |
NM_001204983.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015182.mm.1 |
TC1000002164.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000015184.mm.1 |
NM_001204983.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015184.mm.1 |
NM_001204983.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015184.mm.1 |
TC1000002164.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000015180.mm.1 |
NM_001204983.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015180.mm.1 |
NM_001204983.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015180.mm.1 |
TC1000002164.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000015181.mm.1 |
NM_001204983.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015181.mm.1 |
NM_001204983.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015181.mm.1 |
TC1000002164.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000015183.mm.1 |
NM_001204983.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015183.mm.1 |
NM_001204983.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000015183.mm.1 |
TC1000002164.mm.1 |
MTA TC |