Assay Details
| Gene Symbol: Cep85l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204983.1 | NP_001191912.1 | 3 - 4 | 1293 | 74 | |
| GenBank mRNA | AK039734.1 | - | 3 - 4 | 1295 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cep85l
Genomic Map