Assay Details
| Gene Symbol: Syf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026780.3 | NP_081056.1 | 3 - 4 | 270 | 74 | |
| GenBank mRNA | AK003787.1 | - | 3 - 4 | 272 | 74 | |
| AK152794.1 | - | 3 - 4 | 259 | 74 | ||
| AY033432.1 | - | 3 - 4 | 271 | 74 | ||
| BC051484.1 | - | 3 - 4 | 264 | 1380946 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syf2
Genomic Map