Assay Details
| Gene Symbol: L3mbtl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172787.2 | NP_766375.1 | 4 - 5 | 376 | 71 | |
| GenBank mRNA | AK031398.1 | - | 4 - 5 | 362 | 71 | |
| AK045667.1 | - | 4 - 5 | 377 | 71 | ||
| AK049597.1 | - | 4 - 5 | 411 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

L3mbtl3
Genomic Map