Assay Details
| Gene Symbol: L3mbtl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172787.2 | NP_766375.1 | 9 - 10 | 954 | 122 | |
| GenBank mRNA | AK045667.1 | - | 9 - 10 | 955 | 122 | |
| AK049597.1 | - | 9 - 10 | 989 | 122 | ||
| AK129443.1 | - | 5 - 6 | 499 | 122 | ||
| BC082309.1 | - | 8 - 9 | 824 | 30356672 | 122 | |
| BC085192.1 | - | 8 - 9 | 1059 | 30694487 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

L3mbtl3
Genomic Map