Assay Details
| Gene Symbol: Srrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130477.1 | NP_001123949.1 | 10 - 11 | 1467 | 71 | |
| NM_016799.3 | NP_058079.2 | 10 - 11 | 1482 | 71 | ||
| GenBank mRNA | AF062655.1 | - | 10 - 11 | 1408 | 71 | |
| AK028412.1 | - | 10 - 11 | 1400 | 71 | ||
| BC079542.1 | - | 9 - 10 | 3449 | 71 | ||
| BC094322.1 | - | 10 - 11 | 1397 | 6401250 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Srrm1
Genomic Map