Assay Details
Gene Symbol: Srrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130477.1 | NP_001123949.1 | 2 - 3 | 214 | 116 | |
NM_016799.3 | NP_058079.2 | 2 - 3 | 214 | 116 | ||
GenBank mRNA | AF062655.1 | - | 2 - 3 | 140 | 116 | |
AK008284.1 | - | 2 - 3 | 142 | 116 | ||
AK028412.1 | - | 2 - 3 | 146 | 116 | ||
AK169762.1 | - | 140 | 116 | |||
BC079542.1 | - | 2 - 3 | 134 | 116 | ||
BC094322.1 | - | 2 - 3 | 129 | 6401250 | 116 | |
BU514596.1 | - | 2 - 3 | 214 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
