Assay Details
| Gene Symbol: Ccdc30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270435.1 | NP_001257364.1 | 13 - 14 | 2091 | 92 | |
| NM_001270446.1 | NP_001257375.1 | 8 - 9 | 1347 | 92 | ||
| NM_028506.1 | NP_082782.1 | 9 - 10 | 1354 | 92 | ||
| NM_029286.2 | NP_083562.1 | 13 - 14 | 2106 | 92 | ||
| GenBank mRNA | AK049010.1 | - | 8 - 9 | 1247 | 92 | |
| AK078362.1 | - | 13 - 14 | 2033 | 92 | ||
| AK163185.1 | - | 13 - 14 | 2032 | 92 | ||
| AK224940.1 | - | 9 - 10 | 1283 | 92 | ||
| AK224961.1 | - | 8 - 9 | 1346 | 92 | ||
| BC114436.1 | - | 13 - 14 | 2033 | 40059415 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc30
Genomic Map