Assay Details
| Gene Symbol: Rnf217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146349.1 | NP_001139821.1 | 2 - 3 | 1572 | 71 | |
| GenBank mRNA | AK219068.1 | - | 1 - 2 | 137 | 71 | |
| AK219870.1 | - | 1 - 2 | 36 | 71 | ||
| BF467827.1 | - | 2 - 3 | 357 | 71 | ||
| CN459606.1 | - | 1 - 2 | 114 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf217
Genomic Map