Assay Details
Gene Symbol: Zswim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029912.2 | NP_001025083.1 | 12 - 13 | 2679 | 77 | |
GenBank mRNA | AK122519.1 | - | 12 - 13 | 2679 | 77 | |
BC089375.1 | - | 1 - 2 | 93 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
