| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400027010.mm.1 |
TC0400003368.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027010.mm.1 |
NM_001033774.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027010.mm.1 |
NM_001033774.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027010.mm.1 |
NM_001145637.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027010.mm.1 |
NM_001145637.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027013.mm.1 |
TC0400003368.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027013.mm.1 |
NM_001033774.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027013.mm.1 |
NM_001033774.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027013.mm.1 |
NM_001145637.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027013.mm.1 |
NM_001145637.1 |
Refseq |