Assay Details
| Gene Symbol: Gm1661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033774.2 | NP_001028946.1 | 2 - 3 | 308 | 69 | |
| NM_001145637.1 | NP_001139109.1 | 2 - 3 | 308 | 69 | ||
| GenBank mRNA | AK132996.1 | - | 2 - 3 | 309 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132996.1, NM_001033774.2, NM_001145637.1 transcripts.
Set Membership: |
|

Gm1661
Genomic Map