Assay Details
Gene Symbol: Gm1661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033774.2 | NP_001028946.1 | 3 - 4 | 540 | 71 | |
NM_001145637.1 | NP_001139109.1 | 3 - 4 | 540 | 71 | ||
GenBank mRNA | AK132996.1 | - | 3 - 4 | 541 | 71 | |
BC147060.1 | - | 1 - 2 | 240 | 9056068 | 71 | |
BC147061.1 | - | 1 - 2 | 240 | 9056035 | 71 | |
BC171980.1 | - | 1 - 2 | 240 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
