Assay Details
| Gene Symbol: Gm1661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033774.2 | NP_001028946.1 | 4 - 5 | 602 | 80 | |
| NM_001145637.1 | NP_001139109.1 | 4 - 5 | 602 | 80 | ||
| GenBank mRNA | AK132996.1 | - | 4 - 5 | 603 | 80 | |
| BC147060.1 | - | 2 - 3 | 302 | 9056068 | 80 | |
| BC147061.1 | - | 2 - 3 | 302 | 9056035 | 80 | |
| BC171980.1 | - | 2 - 3 | 302 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gm1661
Genomic Map