Assay Details
| Gene Symbol: Tie1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011587.2 | NP_035717.2 | 3 - 4 | 621 | 67 | |
| GenBank mRNA | AK051512.1 | - | 3 - 4 | 567 | 67 | |
| AK052192.1 | - | 3 - 4 | 541 | 67 | ||
| AK052413.1 | - | 3 - 4 | 554 | 67 | ||
| AK161898.1 | - | 3 - 4 | 541 | 67 | ||
| BC046452.1 | - | 3 - 4 | 543 | 4195879 | 67 | |
| BC057004.1 | - | 3 - 4 | 527 | 6831929 | 67 | |
| BC060182.1 | - | 3 - 4 | 527 | 6831929 | 67 | |
| X71425.1 | - | 3 - 4 | 522 | 67 | ||
| X73960.1 | - | 3 - 4 | 521 | 67 | ||
| X80764.1 | - | 3 - 4 | 835 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tie1
Genomic Map