Assay Details
| Gene Symbol: Kdm4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161823.1 | NP_001155295.1 | 1 - 2 | 148 | 100 | |
| NM_172382.2 | NP_759014.2 | 1 - 2 | 148 | 100 | ||
| GenBank mRNA | AK054095.1 | - | 1 - 2 | 112 | 100 | |
| AK145066.1 | - | 1 - 2 | 134 | 100 | ||
| BC028866.1 | - | 1 - 2 | 51 | 4207552 | 100 | |
| BY308642.1 | - | 1 - 2 | 149 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK054095.1, AK145066.1, BC028866.1, NM_001161823.1, NM_172382.2 transcripts.
Set Membership: |
|

Kdm4a
Genomic Map