Assay Details
| Gene Symbol: Ephb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290753.1 | NP_001277682.1 | 1 - 2 | 244 | 104 | |
| NM_010142.3 | NP_034272.1 | 1 - 2 | 244 | 104 | ||
| GenBank mRNA | BC043088.1 | - | 1 - 2 | 168 | 6418088 | 104 |
| BC062924.1 | - | 1 - 2 | 189 | 104 | ||
| CJ067955.1 | - | 1 - 2 | 244 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC043088.1, NM_001290753.1, NM_010142.3 transcripts.
Set Membership: |
|

Ephb2
Genomic Map